missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-methylpyridine, 97%

Manufacturer:  Alfa Aesar™ A1022436

 View more versions of this product

Catalog No. AAA1022436



Specifications

Specifications

2-Amino-3-methylpyridine
Unpleasant
29°C to 32°C
1603-40-3
MFCD00006320
107892
RGDQRXPEZUNWHX-UHFFFAOYSA-N
3-methylpyridin-2-amine
15347
97%
118°C (244°F)
221°C to 222°C
500g
C6H8N2
UN2811
2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
CC1=C(N=CC=C1)N
108.144
108.14

Chemical Identifiers

1603-40-3
108.144
RGDQRXPEZUNWHX-UHFFFAOYSA-N
15347
CC1=C(N=CC=C1)N
C6H8N2
MFCD00006320
2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
3-methylpyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P307+P311
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed: Call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : US1850000

TSCA : TSCA

SDS Documents