missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5477303

 View more versions of this product

Catalog No. AAH5477303



Description

Description

Applications
It is used as primary and secondary pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Refrigerated (2-4°C). Incompatible with oxidizing agents.
Specifications

Specifications

2-Amino-3-methylpyridine-5-boronic acid pinacol ester
1g
C12H19BN2O2
2-amino-3-methylpyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)C
234.106
234.1
128°C to 130°C
1111637-91-2
MFCD12923388
BWEOUBYDXFBFMA-UHFFFAOYSA-N
3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
53216772
96%

Chemical Identifiers

1111637-91-2
234.106
BWEOUBYDXFBFMA-UHFFFAOYSA-N
53216772
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)C
C12H19BN2O2
MFCD12923388
2-amino-3-methylpyridine-5-boronic acid pinacol ester
3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents