Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-methyl-5-nitropyridine, 97%

$52.03 - $681.91

Chemical Identifiers

Chemical Name or Material 2-Amino-3-methyl-5-nitropyridine
CAS 18344-51-9
Molecular Formula C6H7N3O2
Formula Weight 153.14
MDL Number MFCD03095060
Beilstein 126947
Synonym 2-Amino-5-nitro-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1984403

  SDS
alfa aesar™
L1984403
1g Each for $52.03

AAL1984406

  SDS
alfa aesar™
L1984406
5g Each for $175.47

AAL1984414

  SDS
alfa aesar™
L1984414
25g Each for $681.91
Description & Specifications

Specifications

Formula Weight 153.14
Chemical Name or Material 2-Amino-3-methyl-5-nitropyridine
Molecular Formula C6H7N3O2
CAS 18344-51-9
Synonym 2-Amino-5-nitro-3-picoline
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Melting Point 258° to 261°C
MDL Number MFCD03095060
Quantity 1g
Beilstein 126947
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.