missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-methoxypyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC453740050

 View more versions of this product

Catalog No. AC453740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-3-methoxypyridine
5g
96% min. (GC)
2-amino-3-methoxypyridine, 2-pyridinamine, 3-methoxy, 3-methoxy-2-aminopyridine, 3-methoxy-2-pyridinamine, 3-methoxy-2-pyridylamine, 3-methoxy-pyridin-2-amine, 3-methoxy-pyridin-2-ylamine, 3-methoxypyridine-2-amine, acmc-1c0wi, ksc494m4p
COC1=C(N=CC=C1)N
124.14
124.14
75°C to 77°C
10201-71-5
C6H8N2O
HNAYRVKSWGSQTP-UHFFFAOYSA-N
3-methoxypyridin-2-amine
9920384
97%

Chemical Identifiers

10201-71-5
124.14
2-amino-3-methoxypyridine, 2-pyridinamine, 3-methoxy, 3-methoxy-2-aminopyridine, 3-methoxy-2-pyridinamine, 3-methoxy-2-pyridylamine, 3-methoxy-pyridin-2-amine, 3-methoxy-pyridin-2-ylamine, 3-methoxypyridine-2-amine, acmc-1c0wi, ksc494m4p
3-methoxypyridin-2-amine
C6H8N2O
HNAYRVKSWGSQTP-UHFFFAOYSA-N
9920384
COC1=C(N=CC=C1)N
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents