Please login to your online account to display your discounted pricing

2-Amino-3-methoxypyridine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Amino-3-methoxypyridine
Assay Percent Range 97%
Molecular Formula C6H8N2O
Formula Weight 124.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC453740050

  SDS
thermo fisher scientific
AC453740050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC453740010

  SDS
thermo fisher scientific
AC453740010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Amino-3-methoxypyridine
CAS 10201-71-5
Assay Percent Range 97%
Molecular Formula C6H8N2O
Formula Weight 124.14
Quantity 5g