Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-iodopyridine, 98%

$174.78 - $729.27

Chemical Identifiers

Chemical Name or Material 2-Amino-3-iodopyridine
CAS 104830-06-0
Molecular Formula C5H5IN2
Formula Weight 220.01
MDL Number MFCD01651885
Synonym 3-Iodo-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5000103

  SDS
alfa aesar™
H5000103
1g Each for $174.78

AAH5000106

  SDS
alfa aesar™
H5000106
5g Each for $729.27
Description & Specifications

Specifications

Formula Weight 220.01
CAS 104830-06-0
Melting Point 88° to 92°C
Chemical Name or Material 2-Amino-3-iodopyridine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01651885
Assay 98%
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Synonym 3-Iodo-2-pyridinamine
Quantity 1g
Molecular Formula C5H5IN2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.