missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-3-iodopyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441480010
Specifications
2-Amino-3-iodopyridine | |
Glass bottle | |
89.6°C to 90.6°C | |
104830-06-0 | |
100.0 | |
C5H5IN2 | |
UUDNBWSHTUFGDQ-UHFFFAOYSA-N | |
3-iodopyridin-2-amine | |
2763151 | |
Powder, Crystalline Powder or Needles |
Authentic | |
Brown to White | |
1g | |
96.0 | |
96% min. (GC) | |
2-amino-3-iodopyridine, 2-amino-3-iodpyridine, 2-pyridinamine, 3-iodo, 3-iodo-2-pyridylamine, 3-iodo-pyridin-2-ylamine, 3-iodopyrid-2-ylamine, acmc-20a0df, iodo-aminopyridine, pubchem1265 | |
C1=CC(=C(N=C1)N)I | |
220.01 | |
220.01 | |
97% |
Chemical Identifiers
104830-06-0 | |
220.01 | |
2-amino-3-iodopyridine, 2-amino-3-iodpyridine, 2-pyridinamine, 3-iodo, 3-iodo-2-pyridylamine, 3-iodo-pyridin-2-ylamine, 3-iodopyrid-2-ylamine, acmc-20a0df, iodo-aminopyridine, pubchem1265 | |
3-iodopyridin-2-amine |
C5H5IN2 | |
UUDNBWSHTUFGDQ-UHFFFAOYSA-N | |
2763151 | |
C1=CC(=C(N=C1)N)I |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning