Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-iodo-5-methylpyridine, 95%

$98.55 - $224.51

Chemical Identifiers

Chemical Name or Material 2-Amino-3-iodo-5-methylpyridine
CAS 211308-79-1
Molecular Formula C6H7IN2
Formula Weight 234.04
MDL Number MFCD09037445
Synonym 6-Amino-5-iodo-3-picoline; 3-Iodo-5-methyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2778703

  SDS
alfa aesar™
H2778703
1g Each for $224.51

AAH27787MD

  SDS
alfa aesar™
H27787MD
250mg Each for $98.55
Description & Specifications

Specifications

Formula Weight 234.04
Chemical Name or Material 2-Amino-3-iodo-5-methylpyridine
Assay 95%
Molecular Formula C6H7IN2
Melting Point 86° to 89°C
MDL Number MFCD09037445
CAS 211308-79-1
Quantity 1g
Synonym 6-Amino-5-iodo-3-picoline; 3-Iodo-5-methyl-2-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.