missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-fluorobenzonitrile, 95%

$88.82 - $282.20

Chemical Identifiers

CAS 115661-37-5
Molecular Formula C7H5FN2
Molecular Weight (g/mol) 136.129
InChI Key UNISSOLHERSZOW-UHFFFAOYSA-N
Synonym 2-amino-3-fluorobenzenecarbonitrile
PubChem CID 2783392
IUPAC Name 2-amino-3-fluorobenzonitrile
SMILES C1=CC(=C(C(=C1)F)N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61580MD View Documents Alfa Aesar™
H61580MD
250mg Each for $88.82
 
AAH6158003 View Documents Alfa Aesar™
H6158003
1g Each for $282.20
 
Specifications

Chemical Identifiers

115661-37-5
136.129
2-amino-3-fluorobenzenecarbonitrile
2-amino-3-fluorobenzonitrile
C7H5FN2
UNISSOLHERSZOW-UHFFFAOYSA-N
2783392
C1=CC(=C(C(=C1)F)N)C#N

Specifications

250mg
C7H5FN2
2-amino-3-fluorobenzenecarbonitrile
C1=CC(=C(C(=C1)F)N)C#N
136.129
136.13
95%
115661-37-5
UN3439
UNISSOLHERSZOW-UHFFFAOYSA-N
2-amino-3-fluorobenzonitrile
2783392
Solid
2-Amino-3-fluorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.