missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

CAS 1831130-33-6
Synonym 2-amino-3-fluorobenzeneboronic acid pinacol ester
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62033MD View Documents Alfa Aesar™
H62033MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6203303 View Documents Alfa Aesar™
H6203303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1831130-33-6
2-amino-3-fluorobenzeneboronic acid pinacol ester

Specifications

250mg
2-amino-3-fluorobenzeneboronic acid pinacol ester
Solid
2-Amino-3-fluorobenzeneboronic acid pinacol ester
1831130-33-6
237.08
96%
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures