Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-fluoro-5-methylpyridine, 98%

$90.50 - $361.73

Chemical Identifiers

Chemical Name or Material 2-Amino-3-fluoro-5-methylpyridine
CAS 1211590-31-6
Molecular Formula C6H7FN2
Formula Weight 126.13
MDL Number MFCD13185333
Synonym 2-Amino-3-fluoro-5-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6458303

 
alfa aesar™
H6458303
1g Each for $90.50

AAH6458306

 
alfa aesar™
H6458306
5g Each for $361.73

AAH64583MD

 
alfa aesar™
H64583MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.13
Chemical Name or Material 2-Amino-3-fluoro-5-methylpyridine
Assay 98%
Molecular Formula C6H7FN2
MDL Number MFCD13185333
CAS 1211590-31-6
Synonym 2-Amino-3-fluoro-5-picoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
2-Amino-3-fluoro-5-methylpyridine is a pyridine compound for proteomics research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.