Please login to your online account to display your discounted pricing

2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan
CAS 24386-17-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H8N2O3
Formula Weight 240.22
MDL Number MFCD00099027
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

NRB00495CB

  SDS
fisher scientific company
NRB00495CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.22
Quantity 250mg
Assay 97%
CAS Min % 97.0
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00099027
Packaging Amber Glass Bottle
CAS 24386-17-2
Molecular Formula C13H8N2O3
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.