Please login to your online account to display your discounted pricing

2-Amino-3-cyano-4,5-di(fur-2-yl)furan, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  NRB00495CB

 View more versions of this product

Catalog No. NRB00495CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 240.22
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Molecular Formula C13H8N2O3
MDL Number MFCD00099027
CAS 24386-17-2
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

Specifications

Chemical Name or Material 2-Amino-3-cyano-4, 5-di(fur-2-yl)furan
CAS 24386-17-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H8N2O3
Formula Weight 240.22
MDL Number MFCD00099027
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents