missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-3-chloropyridine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC453710050
Specifications
2-Amino-3-chloropyridine | |
5g | |
96% min. (GC) | |
2-amino-3-chloro pyridine, 2-amino-3-chloro-pyridine, 2-amino-3-chloropyridine, 2-pyridinamine, 3-chloro, 3-chloranylpyridin-2-amine, 3-chloro-2-pyridinamine, 3-chloro-2-pyridinylamine, 3-chloro-2-pyridylamine, 3-chloro-pyridin-2-ylamine, o-aminochloropyridine | |
C1=CC(=C(N=C1)N)Cl | |
128.56 | |
128.56 |
58.0°C to 60.0°C | |
39620-04-7 | |
C5H5ClN2 | |
RZJPBQGRCNJYBU-UHFFFAOYSA-N | |
3-chloropyridin-2-amine | |
693267 | |
97% |
Chemical Identifiers
39620-04-7 | |
128.56 | |
2-amino-3-chloro pyridine, 2-amino-3-chloro-pyridine, 2-amino-3-chloropyridine, 2-pyridinamine, 3-chloro, 3-chloranylpyridin-2-amine, 3-chloro-2-pyridinamine, 3-chloro-2-pyridinylamine, 3-chloro-2-pyridylamine, 3-chloro-pyridin-2-ylamine, o-aminochloropyridine | |
3-chloropyridin-2-amine |
C5H5ClN2 | |
RZJPBQGRCNJYBU-UHFFFAOYSA-N | |
693267 | |
C1=CC(=C(N=C1)N)Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning