Please login to your online account to display your discounted pricing

2-Amino-3-chloropyridine, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Amino-3-chloropyridine
Assay Percent Range 97%
Molecular Formula C5H5ClN2
Formula Weight 128.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC453710010

  SDS
thermo fisher scientific
AC453710010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC453710050

  SDS
thermo fisher scientific
AC453710050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.56
Quantity 1g
Chemical Name or Material 2-Amino-3-chloropyridine
Assay Percent Range 97%
Molecular Formula C5H5ClN2
CAS 39620-04-7