Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-chloropyridine, 95%

$275.53 - $920.49

Chemical Identifiers

Chemical Name or Material 2-Amino-3-chloropyridine
CAS 39620-04-7
Molecular Formula C5H5ClN2
Formula Weight 128.56
MDL Number MFCD03541052
Synonym 3-Chloro-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3016603

 
alfa aesar™
H3016603
1g Each for $275.53

AAH3016606

 
alfa aesar™
H3016606
5g Each for $920.49
Description & Specifications

Specifications

Formula Weight 128.56
Chemical Name or Material 2-Amino-3-chloropyridine
Melting Point 59° to 62°C
Quantity 1g
Assay 95%
Molecular Formula C5H5ClN2
MDL Number MFCD03541052
CAS 39620-04-7
Synonym 3-Chloro-2-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.