Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-chloropyrazine, 97%

$345.05 - $345.05

Chemical Identifiers

Chemical Name or Material 2-Amino-3-chloropyrazine
CAS 6863-73-6
Molecular Formula C4H4ClN3
Formula Weight 129.55
MDL Number MFCD04114305
Synonym 3-Chloro-2-pyrazinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2721506

  SDS
alfa aesar™
H2721506
5g Each for $345.05
Description & Specifications

Specifications

Formula Weight 129.55
Chemical Name or Material 2-Amino-3-chloropyrazine
Assay 97%
Molecular Formula C4H4ClN3
Synonym 3-Chloro-2-pyrazinamine
Health Hazard 1 Warning
CAS 6863-73-6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Melting Point 167° to 171°C
MDL Number MFCD04114305
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
It is employed in the synthesis and tuberculostatic activity of N-pyrazinylthiourea.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.