Please login to your online account to display your discounted pricing

2-Amino-3-chloro-5-nitrobenzonitrile, 98%, Acros Organics

$166.33 - $166.33

Chemical Identifiers

Chemical Name or Material 2-Amino-3-chloro-5-nitrobenzonitrile
Name Note 98%
CAS 20352-84-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4ClN3O2
Formula Weight 197.58
MDL Number MFCD00126957
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC343670250

  SDS
thermo fisher scientific
343670250
Plastic bottle 25g Each for $166.33
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 197.58
Quantity 25g
Color Dark yellow
EINECS Number 243-760-0
TSCA TSCA
Melting Point 180° to 185°C
CAS Max % 100.0
Packaging Plastic bottle
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00126957
CAS Min % 97.5
Chemical Name or Material 2-Amino-3-chloro-5-nitrobenzonitrile
CAS 20352-84-5
Name Note 98%
Molecular Formula C7H4ClN3O2
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.