Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromopyridine, 98%

$43.84 - $174.78

Chemical Identifiers

Chemical Name or Material 2-Amino-3-bromopyridine
CAS 13534-99-1
Molecular Formula C5H5BrN2
Formula Weight 173.01
MDL Number MFCD03095187
Beilstein 109867
Synonym 3-Bromo-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2000603

  SDS
alfa aesar™
L2000603
1g Each for $43.84

AAL2000606

  SDS
alfa aesar™
L2000606
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 173.01
CAS 13534-99-1
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Amino-3-bromopyridine
Health Hazard 1 Warning
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD03095187
Melting Point 64° to 67°C
Synonym 3-Bromo-2-pyridinamine
Molecular Formula C5H5BrN2
Quantity 1g
Beilstein 109867
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.