missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromobenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H6192506

 View more versions of this product

Catalog No. AAH6192506



Description

Description

Applications
Used as pharmaceutical intermediates.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

2-Amino-3-bromobenzoic acid
174°C to 178°C
20776-51-6
MFCD03618453
2-amino-3-bromo benzoic acid
C1=CC(=C(C(=C1)Br)N)C(=O)O
216.034
216.03
97%
White to Yellow
5g
C7H6BrNO2
2936928
SRIZNTFPBWRGPB-UHFFFAOYSA-N
2-amino-3-bromobenzoic acid
270259
Solid

Chemical Identifiers

20776-51-6
216.034
SRIZNTFPBWRGPB-UHFFFAOYSA-N
270259
C1=CC(=C(C(=C1)Br)N)C(=O)O
C7H6BrNO2
MFCD03618453
2-amino-3-bromo benzoic acid
2-amino-3-bromobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents