Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromo-5-(trifluoromethyl)pyridine, 97%

$32.64 - $90.05

Chemical Identifiers

Chemical Name or Material 2-Amino-3-bromo-5-(trifluoromethyl)pyridine
CAS 79456-30-7
Molecular Formula C6H4BrF3N2
Formula Weight 241.01
MDL Number MFCD07375382
Synonym 3-Bromo-5-trifluoromethyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3299103

  SDS
alfa aesar™
H3299103
1g Each for $90.05

AAH32991MD

  SDS
alfa aesar™
H32991MD
250mg Each for $32.64
Description & Specifications

Specifications

Formula Weight 241.01
Melting Point 98° to 101°C
Molecular Formula C6H4BrF3N2
MDL Number MFCD07375382
CAS 79456-30-7
Chemical Name or Material 2-Amino-3-bromo-5-(trifluoromethyl)pyridine
Assay 97%
Quantity 1g
Synonym 3-Bromo-5-trifluoromethyl-2-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.