missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-bromo-5-nitropyridine, 98%, ACROS Organics™

Chemical Identifiers

CAS 15862-31-4
Molecular Formula C5H4BrN3O2
Molecular Weight (g/mol) 218.01
MDL Number MFCD00955628
InChI Key OFXNHXMPRZDIDM-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2-pyridylamine, 2-pyridinamine, 3-bromo-5-nitro, 3-bromo-5-nitro-pyridin-2-ylamine, 3-bromo-5-nitro-pyridin-2-amine, 5-nitro-3-bromo-2-aminopyridine, pubchem1148, acmc-209dib, ksc496a7l, cbi-bb zero/006111
PubChem CID 2734412
IUPAC Name 3-bromo-5-nitropyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC360590010 SDS View Documents Acros Organics
360590010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

15862-31-4
218.01
OFXNHXMPRZDIDM-UHFFFAOYSA-N
2734412
C1=C(C=NC(=C1Br)N)[N+](=O)[O-]
C5H4BrN3O2
MFCD00955628
2-amino-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2-pyridylamine, 2-pyridinamine, 3-bromo-5-nitro, 3-bromo-5-nitro-pyridin-2-ylamine, 3-bromo-5-nitro-pyridin-2-amine, 5-nitro-3-bromo-2-aminopyridine, pubchem1148, acmc-209dib, ksc496a7l, cbi-bb zero/006111
3-bromo-5-nitropyridin-2-amine

Specifications

Authentic
Beige to Brown-Orange
1g
97.5
98%
MFCD00955628
OFXNHXMPRZDIDM-UHFFFAOYSA-N
3-bromo-5-nitropyridin-2-amine
2734412
Powder
2-Amino-3-bromo-5-nitropyridine
Glass bottle
215°C to 219°C
15862-31-4
100.0
C5H4BrN3O2
2-amino-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2-pyridylamine, 2-pyridinamine, 3-bromo-5-nitro, 3-bromo-5-nitro-pyridin-2-ylamine, 3-bromo-5-nitro-pyridin-2-amine, 5-nitro-3-bromo-2-aminopyridine, pubchem1148, acmc-209dib, ksc496a7l, cbi-bb zero/006111
C1=C(C=NC(=C1Br)N)[N+](=O)[O-]
218.01
218.02
≥98.5%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.