missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-bromo-5-nitropyridine, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 360590010

 View more versions of this product

Catalog No. AC360590010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Amino-3-bromo-5-nitropyridine
97.5
Authentic
Powder
98%
Beige to orange-brown
218.02
MFCD00955628
15862-31-4
100.0
Glass bottle
≥98.5%
215° to 219°C
C5H4BrN3O2
1g

Chemical Identifiers

15862-31-4
100.0
C5H4BrN3O2
MFCD00955628
≥98.5%
97.5
98%
218.02
2-Amino-3-bromo-5-nitropyridine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.