Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromo-5-nitrobenzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2-Amino-3-bromo-5-nitrobenzonitrile
CAS 17601-94-4
Molecular Formula C7H4BrN3O2
Formula Weight 242.03
MDL Number MFCD00054185
UN Number UN3439
Synonym 2-Bromo-6-cyano-4-nitroaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2540403

  SDS
alfa aesar™
B2540403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2540406

  SDS
alfa aesar™
B2540406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2540414

  SDS
alfa aesar™
B2540414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.03
Chemical Name or Material 2-Amino-3-bromo-5-nitrobenzonitrile
TSCA TSCA
Assay 98%
Melting Point 186° to 190°C
Molecular Formula C7H4BrN3O2
Quantity 1g
MDL Number MFCD00054185
CAS 17601-94-4
UN Number UN3439
Synonym 2-Bromo-6-cyano-4-nitroaniline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P307+P311
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed: Call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P307+P311
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed: Call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.