Please login to your online account to display your discounted pricing

2-Amino-3-bromo-5-methylpyridine, 98%, ACROS Organics™

$60.10 - $60.10

Chemical Identifiers

Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
CAS 17282-00-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H7BrN2
Formula Weight 187.04
MDL Number MFCD00068231
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC348590250

  SDS
thermo fisher scientific
AC348590250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC348590050

  SDS
thermo fisher scientific
348590050
Glass bottle 5g Each for $60.10

AC348590010

  SDS
thermo fisher scientific
AC348590010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 187.04
Quantity 25g
Color Yellow to beige or brown
Melting Point 70° to 74°C
CAS Max % 100.0
Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6H7BrN2
CAS 17282-00-7
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00068231
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.