Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromo-5-methylpyridine, 98+%

$128.08 - $128.08

Chemical Identifiers

Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
CAS 17282-00-7
Molecular Formula C6H7BrN2
Formula Weight 187.04
MDL Number MFCD00068231
Beilstein 471829
Synonym 6-Amino-5-bromo-3-picoline; 3-Bromo-5-methyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1409006

 
alfa aesar™
A1409006
5g Each for $128.08
Description & Specifications

Specifications

Formula Weight 187.04
Synonym 6-Amino-5-bromo-3-picoline; 3-Bromo-5-methyl-2-pyridinamine
MDL Number MFCD00068231
Quantity 5g
Odor Unpleasant
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C6H7BrN2
Assay ≥98%
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 471829
Melting Point 70° to 74°C
Health Hazard 1 Warning
CAS 17282-00-7
Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.