missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromo-5-methylbenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1544303

 View more versions of this product

Catalog No. AAA1544303



Specifications

Specifications

2-Amino-3-bromo-5-methylbenzoic acid
1g
C8H8BrNO2
3540992
LCMZECCEEOQWLQ-UHFFFAOYSA-N
2-amino-3-bromo-5-methylbenzoic acid
2774400
≥98%
202°C to 206°C
13091-43-5
MFCD00051705
3-bromo-5-methylanthranilic acid, 2-amino-3-bromo-5-methyl-benzoic acid, 6-amino-5-bromo-m-toluic acid, benzoic acid, 2-amino-3-bromo-5-methyl, acmc-20ao5e, 4-amino-3-bromo-5-carboxytoluene, 2-bromo-6-carboxy-4-methylaniline, benzoic acid,2-amino-3-bromo-5-methyl, 2-azanyl-3-bromanyl-5-methyl-benzoic acid, 2-amino-3-bromo-5-methylbenzoic acid
CC1=CC(=C(C(=C1)C(=O)O)N)Br
230.061
230.07

Chemical Identifiers

13091-43-5
230.061
LCMZECCEEOQWLQ-UHFFFAOYSA-N
2774400
CC1=CC(=C(C(=C1)C(=O)O)N)Br
C8H8BrNO2
MFCD00051705
3-bromo-5-methylanthranilic acid, 2-amino-3-bromo-5-methyl-benzoic acid, 6-amino-5-bromo-m-toluic acid, benzoic acid, 2-amino-3-bromo-5-methyl, acmc-20ao5e, 4-amino-3-bromo-5-carboxytoluene, 2-bromo-6-carboxy-4-methylaniline, benzoic acid,2-amino-3-bromo-5-methyl, 2-azanyl-3-bromanyl-5-methyl-benzoic acid, 2-amino-3-bromo-5-methylbenzoic acid
2-amino-3-bromo-5-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents