Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromo-5-fluorobenzoic acid, 95%

Chemical Identifiers

Chemical Name or Material 2-Amino-3-bromo-5-fluorobenzoic acid
CAS 259269-84-6
Formula Weight 234.02
MDL Number MFCD00662509
Beilstein 15583945
Synonym 3-Bromo-5-fluoroanthranilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6662803

 
alfa aesar™
H6662803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66628MD

 
alfa aesar™
H66628MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.02
CAS 259269-84-6
Chemical Name or Material 2-Amino-3-bromo-5-fluorobenzoic acid
Assay 95%
Synonym 3-Bromo-5-fluoroanthranilic acid
MDL Number MFCD00662509
Physical Form Solid
Quantity 1g
Beilstein 15583945