Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-bromo-5-chloropyridine, 95%

$110.00 - $110.00

Chemical Identifiers

Chemical Name or Material 2-Amino-3-bromo-5-chloropyridine
CAS 26163-03-1
Molecular Formula C5H4BrClN2
Formula Weight 207.46
MDL Number MFCD00955627
Synonym 3-Bromo-5-chloro-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2779903

  SDS
alfa aesar™
H2779903
1g Each for $110.00

AAH2779909

  SDS
alfa aesar™
H2779909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Amino-3-bromo-5-chloropyridine
CAS 26163-03-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 95%
Melting Point 83° to 85°C
Molecular Formula C5H4BrClN2
Formula Weight 207.46
Quantity 1g
MDL Number MFCD00955627
Synonym 3-Bromo-5-chloro-2-pyridinamine
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.