missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-benzyloxypyridine, 98.5%, ACROS Organics™

Manufacturer:  Acros Organics 162980050

 View more versions of this product

Catalog No. AC162980050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-3-benzyloxypyridine
24016-03-3
100.0
Glass bottle
98.5%
Brown to Green
MFCD00006316
NMCBWICNRJLKKM-UHFFFAOYSA-N
3-phenylmethoxypyridin-2-amine
90334
Crystalline Powder
98.5%
98.0
Authentic
≥98.0%
94°C to 98°C
C12H12N2O
5g
C1=CC=C(C=C1)COC2=C(N=CC=C2)N
200.241
200.24

Chemical Identifiers

24016-03-3
200.241
NMCBWICNRJLKKM-UHFFFAOYSA-N
C12H12N2O
MFCD00006316
90334
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 245-983-9

TSCA : TSCA

SDS Documents