Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-benzyloxypyridine, 98+%

$54.19 - $180.09

Chemical Identifiers

Chemical Name or Material 2-Amino-3-benzyloxypyridine
CAS 24016-03-3
Molecular Formula C12H12N2O
Formula Weight 200.24
MDL Number MFCD00006316
Beilstein 392674
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1260306

 
alfa aesar™
A1260306
5g Each for $54.19

AAA1260314

 
alfa aesar™
A1260314
25g Each for $180.09
Description & Specifications

Specifications

Formula Weight 200.24
Chemical Name or Material 2-Amino-3-benzyloxypyridine
Assay ≥98%
Melting Point 92° to 94°C
Molecular Formula C12H12N2O
Quantity 5g
MDL Number MFCD00006316
CAS 24016-03-3
Beilstein 392674
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.