missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-benzyloxypyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5434103

 View more versions of this product

Catalog No. AAH5434103



Specifications

Specifications

2-Amino-3-benzyloxypyridine-5-boronic acid pinacol ester
1g
C18H23BN2O3
2-amino-3-benzyloxypyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)OCC3=CC=CC=C3
326.203
326.2
135°C to 137°C
756520-57-7
MFCD13191381
RPQADWBEACHUFT-UHFFFAOYSA-N
3-phenylmethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
21111425
96%

Chemical Identifiers

756520-57-7
326.203
RPQADWBEACHUFT-UHFFFAOYSA-N
21111425
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)OCC3=CC=CC=C3
C18H23BN2O3
MFCD13191381
2-amino-3-benzyloxypyridine-5-boronic acid pinacol ester
3-phenylmethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents