missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-benzyloxy-5-bromopyridine, 96%

Manufacturer:  Alfa Aesar™ H54667MD

 View more versions of this product

Catalog No. AAH54667MD



Specifications

Specifications

2-Amino-3-benzyloxy-5-bromopyridine
250mg
C12H11BrN2O
2-amino-3-benzyloxy-5-bromo-pyridine
C1=CC=C(C=C1)COC2=CC(=CN=C2N)Br
279.137
279.13
92°C to 94°C
754230-78-9
MFCD11109946
ZFNMAJBVWOHSSF-UHFFFAOYSA-N
5-bromo-3-phenylmethoxypyridin-2-amine
20626554
96%

Chemical Identifiers

754230-78-9
279.137
ZFNMAJBVWOHSSF-UHFFFAOYSA-N
20626554
C1=CC=C(C=C1)COC2=CC(=CN=C2N)Br
C12H11BrN2O
MFCD11109946
2-amino-3-benzyloxy-5-bromo-pyridine
5-bromo-3-phenylmethoxypyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents