missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-benzyloxy-5-bromopyridine, 96%

Manufacturer:  Alfa Aesar™ H5466703

 View more versions of this product

Catalog No. AAH5466703



Specifications

Specifications

2-Amino-3-benzyloxy-5-bromopyridine
1g
C12H11BrN2O
2-amino-3-benzyloxy-5-bromo-pyridine
C1=CC=C(C=C1)COC2=CC(=CN=C2N)Br
279.137
279.13
92°C to 94°C
754230-78-9
MFCD11109946
ZFNMAJBVWOHSSF-UHFFFAOYSA-N
5-bromo-3-phenylmethoxypyridin-2-amine
20626554
96%

Chemical Identifiers

754230-78-9
279.137
ZFNMAJBVWOHSSF-UHFFFAOYSA-N
20626554
C1=CC=C(C=C1)COC2=CC(=CN=C2N)Br
C12H11BrN2O
MFCD11109946
2-amino-3-benzyloxy-5-bromo-pyridine
5-bromo-3-phenylmethoxypyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents