Please login to your online account to display your discounted pricing

2-Amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid, 97%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC146745000

 View more versions of this product

Catalog No. AC146745000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 222.22
Quantity 500mg
Color Almost white to beige
Melting Point 280° to 285°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 6-Fluoro-DL-tryptophan
Assay Percent Range 97%
Molecular Formula C11H11FN2O2
MDL Number MFCD00005653
CAS 7730-20-3
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 6-Fluoro-DL-tryptophan
CAS 7730-20-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H11FN2O2
Formula Weight 222.22
MDL Number MFCD00005653