missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,6-difluorobenzamidine dihydrochloride, 97%

Chemical Identifiers

CAS 190011-83-7
Molecular Formula C7H9Cl2F2N3
Molecular Weight (g/mol) 244.067
MDL Number MFCD22683091
InChI Key RBVYKSQOMAYZNS-UHFFFAOYSA-N
Synonym 2-amino-3,6-difluorobenzamidine dihydrochloride, 2-amino-3,6-difluorobenzenecarboximidamide dihydrochloride, 2-amino-3,6-difluorobenzamidine 2hcl, 2-amino-3,6-difluorobenzimidamide dihydrochloride
PubChem CID 67568295
IUPAC Name 2-amino-3,6-difluorobenzenecarboximidamide;dihydrochloride
SMILES C1=CC(=C(C(=C1F)C(=N)N)N)F.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59788MD View Documents Alfa Aesar™
H59788MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5978803 View Documents Alfa Aesar™
H5978803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

190011-83-7
244.067
RBVYKSQOMAYZNS-UHFFFAOYSA-N
67568295
C1=CC(=C(C(=C1F)C(=N)N)N)F.Cl.Cl
C7H9Cl2F2N3
MFCD22683091
2-amino-3,6-difluorobenzamidine dihydrochloride, 2-amino-3,6-difluorobenzenecarboximidamide dihydrochloride, 2-amino-3,6-difluorobenzamidine 2hcl, 2-amino-3,6-difluorobenzimidamide dihydrochloride
2-amino-3,6-difluorobenzenecarboximidamide;dihydrochloride

Specifications

250mg
C7H9Cl2F2N3
2-amino-3,6-difluorobenzamidine dihydrochloride, 2-amino-3,6-difluorobenzenecarboximidamide dihydrochloride, 2-amino-3,6-difluorobenzamidine 2hcl, 2-amino-3,6-difluorobenzimidamide dihydrochloride
C1=CC(=C(C(=C1F)C(=N)N)N)F.Cl.Cl
244.067
244.07
2-Amino-3,6-difluorobenzamidine dihydrochloride
190011-83-7
MFCD22683091
RBVYKSQOMAYZNS-UHFFFAOYSA-N
2-amino-3,6-difluorobenzenecarboximidamide;dihydrochloride
67568295
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning