missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,6-dichlorobenzoic acid, 95%

Manufacturer:  Alfa Aesar™ L1285003

 View more versions of this product

Catalog No. AAL1285003



Description

Description

Applications
2-Amino-3,6-dichlorobenzoic acid is used as a pharmaceutical intermediate and also finds use in chemical research.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, bases and sodium hypochlorite.
Specifications

Specifications

2-Amino-3,6-dichlorobenzoic acid
1g
C7H5Cl2NO2
3266879
MMIREQFMYZQWLU-UHFFFAOYSA-N
2-amino-3,6-dichlorobenzoic acid
604542
95%
150°C to 154°C
3032-32-4
MFCD00052390
benzoic acid, 2-amino-3,6-dichloro, 3,6-dichloroanthranilic acid, 2-amino-3,6-dichlorobenzoicacid, acmc-1afqi, 6-amino-2,5-dichlorobenzoic acid, benzoic acid,2-amino-3,6-dichloro, 2-azanyl-3,6-bis chloranyl benzoic acid
C1=CC(=C(C(=C1Cl)C(=O)O)N)Cl
206.022
206.03

Chemical Identifiers

3032-32-4
206.022
MMIREQFMYZQWLU-UHFFFAOYSA-N
604542
C1=CC(=C(C(=C1Cl)C(=O)O)N)Cl
C7H5Cl2NO2
MFCD00052390
benzoic acid, 2-amino-3,6-dichloro, 3,6-dichloroanthranilic acid, 2-amino-3,6-dichlorobenzoicacid, acmc-1afqi, 6-amino-2,5-dichlorobenzoic acid, benzoic acid,2-amino-3,6-dichloro, 2-azanyl-3,6-bis chloranyl benzoic acid
2-amino-3,6-dichlorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents