Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-diiodopyridine, 95%

$77.15 - $171.77

Chemical Identifiers

Chemical Name or Material 2-Amino-3,5-diiodopyridine
CAS 23597-15-1
Molecular Formula C5H4I2N2
Formula Weight 345.91
MDL Number MFCD02663852
Synonym 3,5-Diiodo-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2751103

  SDS
alfa aesar™
H2751103
1g Each for $171.77

AAH27511MD

  SDS
alfa aesar™
H27511MD
250mg Each for $77.15
Description & Specifications

Specifications

Formula Weight 345.91
Chemical Name or Material 2-Amino-3,5-diiodopyridine
Assay 95%
Quantity 1g
Molecular Formula C5H4I2N2
MDL Number MFCD02663852
CAS 23597-15-1
Synonym 3,5-Diiodo-2-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.