Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-diiodobenzoic acid, 98+%

$77.91 - $1,142.12

Chemical Identifiers

Chemical Name or Material 2-Amino-3,5-diiodobenzoic acid
CAS 609-86-9
Molecular Formula C7H5I2NO2
Formula Weight 388.93
MDL Number MFCD00007682
Beilstein 779429
Synonym 3,5-Diiodoanthranilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1530806

  SDS
alfa aesar™
A1530806
5g Each for $77.91

AAA1530814

  SDS
alfa aesar™
A1530814
25g Each for $342.04

AAA1530822

  SDS
alfa aesar™
A1530822
100g Each for $1,142.12
Description & Specifications

Specifications

Formula Weight 388.93
CAS 609-86-9
RTECS Number CB2999300
Molecular Formula C7H5I2NO2
TSCA TSCA
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Amino-3,5-diiodobenzoic acid
Melting Point ≈240°C (dec.)
Assay ≥98%
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
MDL Number MFCD00007682
Synonym 3,5-Diiodoanthranilic acid
Beilstein 779429
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.