missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-diiodobenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1530822

 View more versions of this product

Catalog No. AAA1530822



Specifications

Specifications

2-Amino-3,5-diiodobenzoic acid
100g
C7H5I2NO2
779429
RFIBDMCPIREZKC-UHFFFAOYSA-N
2-amino-3,5-diiodobenzoic acid
69116
≥98%
~240°C (decomposition)
609-86-9
MFCD00007682
3,5-diiodoanthranilic acid, benzoic acid, 2-amino-3,5-diiodo, anthranilic acid, 3,5-diiodo, 2-amino-3,5-diiodobenzoicacid, acmc-209mnv, 3,5-diiodoanthranilicacid, 3,5-diiodoanthanilic acid, anthranilic acid,5-diiodo, 4-14-00-01085 beilstein handbook reference, 2-amino-3,5-diiodo-benzoic acid
C1=C(C=C(C(=C1I)N)C(=O)O)I
388.931
388.93

Chemical Identifiers

609-86-9
388.931
RFIBDMCPIREZKC-UHFFFAOYSA-N
69116
C1=C(C=C(C(=C1I)N)C(=O)O)I
C7H5I2NO2
MFCD00007682
3,5-diiodoanthranilic acid, benzoic acid, 2-amino-3,5-diiodo, anthranilic acid, 3,5-diiodo, 2-amino-3,5-diiodobenzoicacid, acmc-209mnv, 3,5-diiodoanthranilicacid, 3,5-diiodoanthanilic acid, anthranilic acid,5-diiodo, 4-14-00-01085 beilstein handbook reference, 2-amino-3,5-diiodo-benzoic acid
2-amino-3,5-diiodobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CB2999300

TSCA : TSCA

Documents