missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-dichlorobenzoic acid, 97%

$66.43 - $987.31

Chemical Identifiers

CAS 2789-92-6
Molecular Formula C7H5Cl2NO2
Formula Weight 206.03
MDL Number MFCD00017088
Beilstein 779100
Synonym 3,5-Dichloroanthranilic acid
Chemical Name or Material 2-Amino-3,5-dichlorobenzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1696909 SDS Alfa Aesar™
A1696909
10g Each for $66.43
AAA1696918 SDS Alfa Aesar™
A1696918
50g Each for $224.39
AAA1696930 SDS Alfa Aesar™
A1696930
250g Each for $987.31
Specifications

Description

Applications
2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
This material is stable if stored under proper conditions. Avoid excessive heat and light. Keep away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

2789-92-6
206.03
779100
2-Amino-3,5-dichlorobenzoic acid
C7H5Cl2NO2
MFCD00017088
3,5-Dichloroanthranilic acid

Specifications

2789-92-6
10g
206.03
779100
2-Amino-3,5-dichlorobenzoic acid
225° to 227°C
C7H5Cl2NO2
MFCD00017088
3,5-Dichloroanthranilic acid
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CB2800000