Learn More
2-Amino-3,5-dichlorobenzoic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1696930
Description
2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-3,5-dichlorobenzoic acid, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
This material is stable if stored under proper conditions. Avoid excessive heat and light. Keep away from strong oxidizing agents.
Specifications
2-Amino-3,5-dichlorobenzoic acid | |
225°C to 227°C | |
MFCD00017088 | |
779100 | |
Insoluble in water. | |
NC1=C(Cl)C=C(Cl)C=C1C(O)=O | |
206.02 | |
206.03 |
2789-92-6 | |
C7H5Cl2NO2 | |
250 g | |
3,5-dichloroanthranilic acid, benzoic acid, 2-amino-3,5-dichloro, anthranilic acid, 3,5-dichloro, 3,5-dichloro-2-aminobenzoic acid, 2-amino-3,5-dichloro-benzoic acid, timtec-bb sbb003493, 2-amino-3,5-dichlorobenzoicacid, acmc-209gzl, rarechem al bo 0770, buttpark 49\07-46 | |
KTHTXLUIEAIGCD-UHFFFAOYSA-N | |
2-amino-3,5-dichlorobenzoic acid | |
76036 | |
97% |
Chemical Identifiers
2789-92-6 | |
206.02 | |
KTHTXLUIEAIGCD-UHFFFAOYSA-N | |
76036 | |
NC1=C(Cl)C=C(Cl)C=C1C(O)=O |
C7H5Cl2NO2 | |
MFCD00017088 | |
3,5-dichloroanthranilic acid, benzoic acid, 2-amino-3,5-dichloro, anthranilic acid, 3,5-dichloro, 3,5-dichloro-2-aminobenzoic acid, 2-amino-3,5-dichloro-benzoic acid, timtec-bb sbb003493, 2-amino-3,5-dichlorobenzoicacid, acmc-209gzl, rarechem al bo 0770, buttpark 49\07-46 | |
2-amino-3,5-dichlorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-520-3
RTECSNumber : CB2800000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only