missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-dibromopyrazine, 99%

$47.57 - $119.15

Chemical Identifiers

CAS 24241-18-7
Molecular Formula C4H3Br2N3
Formula Weight 252.9
MDL Number MFCD00673150
Synonym 3,5-Dibromo-2-pyrazinamine
Chemical Name or Material 2-Amino-3,5-dibromopyrazine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH28744MD SDS Alfa Aesar™
H28744MD
250mg Each for $47.57
AAH2874403 SDS Alfa Aesar™
H2874403
1g Each for $119.15
Specifications Description & Specifications

Chemical Identifiers

24241-18-7
252.9
3,5-Dibromo-2-pyrazinamine
C4H3Br2N3
MFCD00673150
2-Amino-3,5-dibromopyrazine

Specifications

24241-18-7
250mg
252.9
3,5-Dibromo-2-pyrazinamine
99%
114° to 117°C
C4H3Br2N3
MFCD00673150
2-Amino-3,5-dibromopyrazine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning