Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-dibromobenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2-Amino-3,5-dibromobenzonitrile
CAS 68385-95-5
Formula Weight 275.93
MDL Number MFCD00179571
Beilstein 2834670
Synonym 2,4-Dibromo-6-cyanoaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6415503

 
alfa aesar™
H6415503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6415506

 
alfa aesar™
H6415506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 152° to 156°C (lit.)
Formula Weight 275.93
Chemical Name or Material 2-Amino-3,5-dibromobenzonitrile
Physical Form Solid
Assay 97%
Quantity 1g
MDL Number MFCD00179571
CAS 68385-95-5
Beilstein 2834670
Synonym 2,4-Dibromo-6-cyanoaniline