Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-dibromobenzoic acid, 98%

$47.09 - $47.09

Chemical Identifiers

Chemical Name or Material 2-Amino-3,5-dibromobenzoic acid
CAS 609-85-8
Molecular Formula C7H5Br2NO2
Formula Weight 294.93
MDL Number MFCD00017087
Beilstein 779291
Synonym 3,5-Dibromoanthranilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0741906

 
alfa aesar™
L0741906
5g Each for $47.09
Description & Specifications

Specifications

Formula Weight 294.93
Molecular Formula C7H5Br2NO2
CAS 609-85-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Amino-3,5-dibromobenzoic acid
Melting Point 230° to 235°C
Quantity 5g
MDL Number MFCD00017087
Beilstein 779291
Synonym 3,5-Dibromoanthranilic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.