missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-dibromobenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H5601514

 View more versions of this product

Catalog No. AAH5601514



Specifications

Specifications

2-Amino-3,5-dibromobenzaldehyde
130°C to 135°C
50910-55-9
MFCD00671100
RCPAZWISSAVDEA-UHFFFAOYSA-N
2-amino-3,5-dibromobenzaldehyde
688305
97%
Air Sensitive
25g
C7H5Br2NO
2-amino-3,5-dibromo-benzaldehyde
C1=C(C=C(C(=C1Br)N)C=O)Br
278.931
278.93

Chemical Identifiers

50910-55-9
278.931
RCPAZWISSAVDEA-UHFFFAOYSA-N
688305
C1=C(C=C(C(=C1Br)N)C=O)Br
C7H5Br2NO
MFCD00671100
2-amino-3,5-dibromo-benzaldehyde
2-amino-3,5-dibromobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 256-841-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents