Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3,5-dibromo-6-methylpyridine, 97%

$70.07 - $216.97

Chemical Identifiers

Chemical Name or Material 2-Amino-3,5-dibromo-6-methylpyridine
CAS 91872-10-5
Molecular Formula C6H6Br2N2
Formula Weight 265.95
MDL Number MFCD00068229
Beilstein 121839
Synonym 6-Amino-3,5-dibromo-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049803

  SDS
alfa aesar™
B2049803
1g Each for $70.07

AAB2049806

  SDS
alfa aesar™
B2049806
5g Each for $216.97
Description & Specifications

Specifications

Formula Weight 265.95
Chemical Name or Material 2-Amino-3,5-dibromo-6-methylpyridine
Melting Point 144° to 145°C
Assay 97%
Quantity 1g
Molecular Formula C6H6Br2N2
MDL Number MFCD00068229
CAS 91872-10-5
Beilstein 121839
Synonym 6-Amino-3,5-dibromo-2-picoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.