Please login to your online account to display your discounted pricing

2-Amino-3,3-dimethylbutane, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  353000010

 View more versions of this product

Catalog No. AC353000010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 101.19
Quantity 1g
Color Clear colorless to very slightly yellow
EINECS Number 223-353-4
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 2-Amino-3, 3-dimethylbutane
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H15N
Linear Formula (CH3)3CCH(CH3)NH2
MDL Number MFCD00008078
CAS 3850-30-4
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Amino-3, 3-dimethylbutane
Name Note 98%
CAS 3850-30-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H15N
Linear Formula (CH3)3CCH(CH3)NH2
Formula Weight 101.19
MDL Number MFCD00008078
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents