Please login to your online account to display your discounted pricing

2-Amino-3,3-dimethylbutane, 98%, ACROS Organics™

$44.23 - $44.23

Chemical Identifiers

Chemical Name or Material 2-Amino-3, 3-dimethylbutane
Name Note 98%
CAS 3850-30-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H15N
Linear Formula (CH3)3CCH(CH3)NH2
Formula Weight 101.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC353000050

  SDS
thermo fisher scientific
353000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC353000010

  SDS
thermo fisher scientific
353000010
Glass bottle 1g Each for $44.23
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 101.19
Quantity 5g
Color Clear colorless to very slightly yellow
EINECS Number 223-353-4
TSCA TSCA
CAS Max % 100.0
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Name Note 98%
Linear Formula (CH3)3CCH(CH3)NH2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2-Amino-3, 3-dimethylbutane
Molecular Formula C6H15N
CAS Min % 97.5
MDL Number MFCD00008078
Assay Percent Range 98%
CAS 3850-30-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.