Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-2-thiazoline, 97%

$70.35 - $204.73

Chemical Identifiers

Chemical Name or Material 2-Amino-2-thiazoline
CAS 1779-81-3
Molecular Formula C3H6N2S
Formula Weight 102.16
MDL Number MFCD00005313
Beilstein 105367
Synonym 4,5-Dihydro-2-thiazolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0201814

  SDS
alfa aesar™
L0201814
25g Each for $70.35

AAL0201822

  SDS
alfa aesar™
L0201822
100g Each for $204.73
Description & Specifications

Specifications

Formula Weight 102.16
CAS 1779-81-3
Chemical Name or Material 2-Amino-2-thiazoline
Assay 97%
Molecular Formula C3H6N2S
MDL Number MFCD00005313
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Melting Point 77° to 82°C
Beilstein 105367
Synonym 4,5-Dihydro-2-thiazolamine
RTECS Number XJ6540000

Applications
2-Amino-2-thiazoline is used as radiation protectants.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.